EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Zn-mediated electrochemical allylation of aldehydes in aqueous ammonia.
PMID 19662260
Abstract
An efficient electrosynthesis of homoallylic alcohols from allylic bromides and aldehydes in aqueous ammonia is achieved in an undivided cell fitted with a pair of zinc electrodes.