EMAIL THIS PAGE TO A FRIEND
Natural product research
New antitumour fungal metabolites from Alternaria porri.
PMID 19662571
Abstract
Chemical investigation of the onion pathogenic fungus Alternaria porri resulted in the isolation of two new phthalides named zinnimide (2) and deprenylzinnimide (8), along with a new bianthraquinone, alterporriol F (10). The structures of the new metabolites were characterised by spectroscopic analysis and chemical degradation. Of the new compounds isolated, alterporriol F was highly cytotoxic towards HeLa and KB cells, with IC(50) values of 6.5 and 7.0 microg mL(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
35758
Alternariol, analytical standard
C14H10O5
A1312
Alternariol from Alternaria sp., ~96%
C14H10O5