EMAIL THIS PAGE TO A FRIEND
Journal of agricultural and food chemistry
Taste contribution of (R)-strombine to dried scallop.
PMID 19663385
Abstract
A broth of dried scallop adductor muscles was prepared. Tasters appreciated the typical seafood, sweet, slightly umami taste of scallop, which is difficult to reproduce with common ingredients. Therefore, the broth was fractionated and, guided by multiple tastings, we isolated a sweet, umami, delicious fraction. This fraction contained glycine, alanine, and (R)-strombine ((R)-2-(carboxymethylamino)propanoic acid). (R) and (S)-strombine were prepared, and a sensory analysis with 47 judges demonstrated that the taste thresholds were 0.5 g/L for (R)-strombine and 0.7 g/L for (S)-strombine. The sensory attributes were described as salty and umami.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M51008
Methyliminodiacetic acid, 99%
C5H9NO4