EMAIL THIS PAGE TO A FRIEND
Organic letters
Rh-catalyzed reductive coupling reaction of aldehydes with conjugated dienes promoted by triethylborane.
PMID 19663391
Abstract
Rh(I) catalyzes the reductive coupling reaction of a wide variety of aldehydes with conjugated dienes in the presence of a stoichiometric amount of triethylborane to provide homoallyl alcohols in a single operation.