EMAIL THIS PAGE TO A FRIEND
Organic letters
Highly efficient synthesis of DNA-binding hairpin polyamides via the use of a new triphosgene coupling strategy.
PMID 19670849
Abstract
A facile and highly efficient solid phase synthesis method is reported for the preparation of hairpin DNA-binding polyamides using the cost-effective triphosgene (BTC) activating agent. Difficult polyamide sequences were prepared from N-methylimidazole (Im) and N-methylpyrrole (Py) building blocks with high stepwise yields (>98%) using Boc chemistry. The versatility of the triphosgene coupling approach was also demonstrated for the first time on aryl hydrazine resins to afford biomedically relevant tail-truncated polyamides in excellent isolated yields.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M78801
N-Methylpyrrole, 99%
C5H7N