EMAIL THIS PAGE TO A FRIEND
Organic letters
Phosphine-catalyzed [4 + 3] annulation for the synthesis of highly functionalized bicyclo[3.2.2]nonadienes.
PMID 19670854
Abstract
A phosphine-catalyzed [4 + 3] annulation of modified allylic carbonates with methyl coumalate was developed. This strategy offered a powerful method for the construction of bicyclo[3.2.2]nonadiene skeleton with high stereoselectivity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261432
Methyl coumalate, 98%
C7H6O4