EMAIL THIS PAGE TO A FRIEND
Pediatric annals
Treatment of alopecia areata in children.
PMID 19685659
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
339792
3,4-Dibutoxy-3-cyclobutene-1,2-dione, 98%
C12H18O4