EMAIL THIS PAGE TO A FRIEND
Ideggyogyaszati szemle
Current medical aspects of pantethine.
PMID 19685700
Abstract
Pantethine, the stable disulfide form of pantetheine, is the major precursor of coenzyme A, which plays a central role in the metabolism of lipids and carbohydrates. Coenzyme A is a cofactor in over 70 enzymatic pathways, including fatty acid oxidation, carbohydrate metabolism, pyruvate degradation, amino acid catabolism, haem synthesis, acetylcholine synthesis, phase II detoxification, acetylation, etc. Pantethine has beneficial effects in vascular disease, it able to decrease the hyperlipidaemia, moderate the platelet function and prevent the lipid-peroxidation. Moreover its neuro-endocrinological regulating role, its good influence on cataract and cystinosis are also proved. This molecule is a well-tolerated therapeutic agent; the frequency of its side-effect is very low and mild. Based on these preclinical and clinical data, it could be recommended using this compound as adjuvant therapy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P2125
D-Pantethine, analytical standard
C22H42N4O8S2