EMAIL THIS PAGE TO A FRIEND
Critical care medicine
Levosimendan is superior to enoximone in refractory cardiogenic shock complicating acute myocardial infarction.
PMID 19687655
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E1279
Enoximone, solid
C12H12N2O2S