EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Biomimetic lipoglycopolymer membranes: photochemical surface attachment of supramolecular architectures with defined orientation.
PMID 19688795
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
42358
Dioctadecylamine, ≥99.0% (NT)
C36H75N