EMAIL THIS PAGE TO A FRIEND
Organic letters
Synthesis of enol lactones via Cu(I)-catalyzed intramolecular O-vinylation of carboxylic acids.
PMID 19689116
Abstract
With the catalysis of CuI/trans-N,N'-dimethylcyclohexane-1,2-diamine, a number of carboxylic acids underwent efficient intramolecular O-vinylation with vinyl bromides leading to the synthesis of the corresponding five- and six-membered enol lactones. The same catalytic system also led to the efficient cycloisomerization of alkynoic acids.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
V1902
Vinyl bromide, 98%
C2H3Br
434191
Vinyl bromide solution, 1.0 M in THF
C2H3Br