EMAIL THIS PAGE TO A FRIEND
Mini reviews in medicinal chemistry
The use of intravenous aminobisphosphonates for the treatment of Paget's disease of bone.
PMID 19689402
Abstract
Paget's disease of bone is a focal skeletal disorder characterized by the formation of structurally abnormal bone, deformity and other complications leading to significant disability and bone pain. Recently, the availability of newer, more potent nitrogen-containing bisphosphonates has improved treatment outcomes, allowing a more effective and convenient management of this disorder.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N6037
Neridronate, ≥97% (NMR), solid
C6H17NO7P2