EMAIL THIS PAGE TO A FRIEND
Journal of hepatology
Acute hepatitis due to brivudin: a case report.
PMID 19692139
Abstract
Brivudin is licensed in several European countries for the treatment of herpetic infections, and is considered safe (approximately 1% of patients with transient elevation of liver enzymes) in large multicenter trials. We report a case of acute brivudin hepatitis documented with a liver biopsy in detail. Liver biopsy demonstrated acute liver injury with a predominant cytolytic pattern and features suggestive of a drug-induced immunoallergic hepatitis. Elevated ALT levels returned to normal within weeks. This is the first published case of acute immunoallergic hepatitis due to brivudin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B9647
(E)-5-(2-Bromovinyl)-2′-deoxyuridine
C11H13BrN2O5