EMAIL THIS PAGE TO A FRIEND
Photomedicine and laser surgery
Effect of low-energy gallium-aluminum-arsenide and aluminium gallium indium phosphide laser irradiation on the viability of C2C12 myoblasts in a muscle injury model.
PMID 19698002
Abstract
To evaluate the effect of phototherapy on the viability of cultured C2C12 myoblasts under different nutritional conditions (muscle injury model) using low-energy gallium-aluminum-arsenide (GaAlAs) and aluminium-gallium-indium-phosphide (InGaAlP) lasers with different wavelengths and powers. The beneficial effects of phototherapy using low-energy lasers depend on irradiation parameters and type of laser used, but there are no data in the literature on C2C12 myoblasts proliferation after phototherapy with GaAlAs and InGaAlP lasers. A C2C12 cell line cultured in regular (10% fetal bovine serum, FBS) and nutrient-deficient (5% FBS) media were irradiated with low-energy GaAlAs (660 nm) and InGaAlP (780 nm) lasers with energy densities of 3.8, 6.3, and 10 J/cm2, and 3.8, 10, and 17.5 J/cm2, respectively. Cell proliferation was assessed indirectly 24 h after irradiation by measuring the mitochondrial activity and using the crystal violet assay. There were no significant differences in cell viability between laser-treated myoblasts and control cultures for all tested parameters after 24 h of cell culture, although cell cultures grown in regular nutrient medium supplemented with 10% FBS exhibited higher growth rates than cultures, irradiated or not, grown in nutrient-deficient medium. Laser phototherapy did not improve C2C12 viability under regular or nutrient-deficient (muscle injury model) conditions using the above parameters.