EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
Synthesis of thionato(triethylphosphine) gold(I) complexes:analogues of "auranofin," an antiarthritic drug.
PMID 1969932
Abstract
New compounds having the formula [L-Au-PEt3]Cl (where L = imidazolidine-2-thione(Imt) and 1,3-Diazinane-2-thione(Diaz)) are synthesized. They are characterized by elemental analysis, infrared, 13C and 31P NMR spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
288225
Chloro(triethylphosphine)gold(I), 97%
C6H15AuClP