EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
Lithium phenolates solvated by tetrahydrofuran and 1,2-dimethoxyethane: structure determination using the method of continuous variation.
PMID 19702308
Abstract
The method of continuous variation in conjunction with (6)Li NMR spectroscopy was used to characterize lithium phenolates solvated by tetrahydrofuran and 1,2-dimethoxyethane. The strategy relies on the formation of ensembles of homo- and heteroaggregated phenolates. The symmetries and concentration dependencies of the heteroaggregates attest to the aggregation numbers of the homoaggregates. The structurally diverse phenols afford substrate- and solvent-dependent combinations of lithium phenolate monomers, dimers, trimers, tetramers, and pentamers. We discuss the refinement of protocols for characterizing O-lithiated species. Computational studies examine further the substituent and solvent dependencies of aggregation.