EMAIL THIS PAGE TO A FRIEND
Anesthesiology
General anesthesia and ascending arousal pathways.
PMID 19707121
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T6764
Triprolidine hydrochloride, ≥99%
C19H22N2 · HCl