EMAIL THIS PAGE TO A FRIEND
Journal of neuroscience methods
Combination of diOlistic labeling with retrograde tract tracing and immunohistochemistry.
PMID 19712695
Abstract
Neuronal staining techniques have played a crucial role in the analysis of neuronal function. Several different staining techniques have been developed to allow morphological analyses of neurons. DiOlistic labeling, in which beads are coated with a lipophilic dye and then ballistically ejected onto brain tissue, has recently been introduced as a useful and simple means to label neurons and glia in their entirety. Although diOlistic labeling provides detailed information on the morphology of neurons, combining this approach with other staining methods is a significant advance. We have developed protocols that result in high quality diOlistically- and retrogradely-labeled or diOlistically-immunohistochemically labeled neurons. These dual-label methods require modification of fixation parameters and the restricted use of detergents for tissue permeabilization, and are readily applicable to a wide range of tracers and antibodies.