EMAIL THIS PAGE TO A FRIEND
Clinical journal of the American Society of Nephrology : CJASN
Sodium bicarbonate for the prevention of contrast induced-acute kidney injury: a systematic review and meta-analysis.
PMID 19713291