EMAIL THIS PAGE TO A FRIEND
Molecular pharmacology
Functional characterization of human cytochrome P450 2S1 using a synthetic gene-expressed protein in Escherichia coli.
PMID 19713358
Abstract
Human cytochrome P450 2S1 was recently identified and shown to be inducible by 2,3,7,8-tetrachlorodibenzo-p-dioxin and hypoxia. It is highly expressed in epithelial cells of tissues that are exposed to the environment and in many tumors of epithelial origin. The biological function of CYP2S1 has not yet been determined, although its possible role in carcinogen metabolism has been suggested. In this report, we investigated its ability to metabolize carcinogens. To obtain a large quantity of active enzyme for substrate screening, we overexpressed CYP2S1 in Escherichia coli (200 nM culture), using a synthetic gene approach. High-level expression allowed us to achieve purification of CYP2S1 with high specific content and purity (16 nmol/mg). Despite high-level expression, we found that CYP2S1 was not readily reduced by cytochrome P450 reductase, and thus no activity was found using NADPH. However, the oxidative activity of CYP2S1 was supported by cumene hydroperoxide or H(2)O(2), such that CYP2S1 oxidized many important environmental carcinogens, including benzo[a]pyrene, 9,10-dihydro-benzo[a]pyrene, 7,12-dimethylbenz[a]anthracene, benzo[a]pyrene-7,8-dihydrodiol, aflatoxin B1, naphthalene, and styrene, with high turnover. Most substrates tested were converted to detoxification products, except in the case of benzo[a]pyrene-7,8-dihydrodiol, which was converted into the very potent carcinogenic metabolite 7,8-dihydrodiol-trans-9,10-epoxide at a relatively efficient rate (K(m) = 12.4 +/- 2 microM, turnover = 2.3 min(-1)). This metabolite formation was also supported both in vitro and in vivo by fatty acid hydroperoxides described in the accompanying report (p. 1044). Together, these data indicate that CYP2S1 contributes to the metabolism of environmental carcinogens via an NADPH independent activity.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
45993
Styrene, analytical standard
C8H8