EMAIL THIS PAGE TO A FRIEND
Journal of chromatography. A
Gas chromatography-mass spectrometry resolution of sugar acid enantiomers on a permethylated beta-cyclodextrin stationary phase.
PMID 19716135
Abstract
Analysis of compounds in meteorites revealed a need to simultaneously characterize multiple enantiomers of sugar acids (aldonic acids) present in trace amounts. Analyses by gas chromatography-mass spectrometry demonstrated that all but two of the three-carbon through six-carbon straight-chained sugar acid enantiomer pairs could be resolved using a single derivatization procedure and one set of GC parameters. Compounds were analyzed as their ethyl ester/O-triflouroacetyl, isopropyl ester/O-triflouroacetyl and isopropyl ester/O-pentafluoropropionyl derivatives on a capillary column containing permethylated beta-cyclodextrin (Chirasil-Dex CB) as the stationary phase. Characteristic mass fragments are related to the ester groups while several ions are also common to derivatized monosaccharides.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
00033
D-Arabinonic acid sodium salt, ≥98.0% (TLC)
C5H9NaO6
09154
L-Xylonic acid lithium salt, ≥95.0% (TLC)
C5H10O6 · xLi+