EMAIL THIS PAGE TO A FRIEND
The journal of physical chemistry. B
Photodegradation mechanism of nonsteroidal anti-inflammatory drugs containing thiophene moieties: suprofen and tiaprofenic acid.
PMID 19719267
Abstract
The photodegradation of nonsteroid anti-inflammatory drugs suprofen, 2-[4-(2-thienoyl)phenyl]propionic acid, and tiaprofenic acid, 2-(5-benzoyl-2-thienyl)propanoic acid, is studied by means of density functional theory. Besides the redox properties of the neutral species, we report on absorption spectra and degradation pathways involving excitation, intersystem crossing to the T(1) state, and spontaneous decarboxylation of the deprotonated species of each drug. The energetics and properties of the suprofen and tiaprofenic acid systems are found to be very similar to those of the highly photolabile benzyl analogue ketoprofen. Mechanisms leading to the formation of a closed-shell decarboxylated ethyl species, as well as peroxyl radicals capable of initiating lipid peroxidation reactions, are discussed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T1410900 Tiaprofenic acid, European Pharmacopoeia (EP) Reference Standard
C14H12O3S