EMAIL THIS PAGE TO A FRIEND
Biological & pharmaceutical bulletin
Glycogen phosphorylase a inhibitors with a phenethylphenylphthalimide skeleton derived from thalidomide-related alpha-glucosidase inhibitors and liver X receptor antagonists.
PMID 19721243
Abstract
Novel glycogen phosphorylase a (GPa) inhibitors with a phenethylphenylphthalimide skeleton were prepared based on alpha-glucosidase inhibitors and liver X receptor (LXR) antagonists derived from thalidomide. Their structure-activity relationships were analyzed. Some of the compounds thus prepared showed potent inhibitory activity against rabbit muscle GPa with more than 10-fold greater efficacy than a typical GPa inhibitor, 1,4-dideoxy-1,4-imino-D-arabinitol.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
416274
N-Phenylphthalimide, 98%
C14H9NO2