EMAIL THIS PAGE TO A FRIEND
Journal of food science
High-temperature natural antioxidant improves soy oil for frying.
PMID 19723188
Abstract
The objectives of this study were to determine the frying stability of soybean oil (SBO) treated with a natural citric acid-based antioxidant, EPT-OILShield able to withstand high temperatures and to establish the oxidative stability of food fried in the treated oil. Soybean oil with 0.05% and 0.5% EPT-OILShield and an untreated control SBO were used for intermittent batch frying of tortilla chips at 180 degrees C for up to 65 h. Oil frying stability was measured by free fatty acids (FFA) and total polar compounds (TPC). Chips were aged for up to 4 mo at 25 degrees C and evaluated for rancid flavor by a 15-member, trained, experienced analytical sensory panel and for hexanal content as an indicator of oxidation. Oil with 0.05% EPT-OILShield had significantly less FFA and TPC than the control. The effect of EPT-OILShield was apparently retained in aged chips because hexanal levels were significantly lower in chips fried in oil with 0.05% EPT-OILShield than in chips fried in the control. Tortilla chips fried in the control were rancid after 2 mo at 25 degrees C at sampling times evaluated from 25 to 65 h; however, chips fried in oil with 0.05% EPT-OILShield and used for 65 h were described as only slightly rancid after 4 mo. Gamma tocopherol levels were significantly higher in the chips fried in the oil with 0.05% EPT-OILShield than in the control, helping to inhibit oxidation in the tortilla chips during storage.