EMAIL THIS PAGE TO A FRIEND
Journal of vector borne diseases
Efficacy of Cyphenothrin (Gokilaht-S 5% EC) as space spray against mosquitoes in sentinel cages.
PMID 19724090
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
46037
Cyphenothrin, PESTANAL®, analytical standard
C24H25NO3