EMAIL THIS PAGE TO A FRIEND
International Journal of Molecular Medicine
Effect of insulin levels on the phosphorylation of specific amino acid residues in IRS-1: implications for burn-induced insulin resistance.
PMID 19724894