EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Enantioselective total synthesis of the marine toxin (-)-gymnodimine employing a Barbier-type macrocyclization.
PMID 19728353