EMAIL THIS PAGE TO A FRIEND
Acta pharmacologica Sinica
Epinephrine, phenylephrine, and methoxamine induce infiltrative anesthesia via alpha1-adrenoceptors in rats.
PMID 19730427
Abstract
To assess whether epinephrine, phenylephrine, and methoxamine act via certain subtypes of adrenoceptors to exert their local anesthetic activity. We investigated cutaneous anesthesia from adrenoceptor agonists and/or antagonists in conscious, unanesthetized Sprague-Dawley male rats (weight 200-250 g). Cutaneous anesthesia was evidenced by a block of the cutaneous trunci muscle reflex, which is characterized by reflex movement of the skin over the back produced by twitches of lateral thoracispinal muscles in response to local dorsal cutaneous noxious pinprick. Local infiltration of epinephrine, L-phenylephrine, or methoxamine alone induces cutaneous anesthesia in rats in a dose-dependent way. Epinephrine is found to be 19 and 29 times more potent than those of methoxamine and L-phenylephrine, respectively. The cutaneous anesthesia induced by epinephrine, phenylephrine, or methoxamine can be significantly reduced by alpha(1)-adrenoceptor antagonists (eg, prazosin), alpha1, alpha2-adrenoceptor antagonist, alpha(1A)-adrenoceptor antagonist (eg, 5-methylurapdil), alpha(1B)-adrenoceptor antagonist (eg, chloroethylclonidine), or alpha(1D)-adrenoceptor antagonist (eg, BMY7873). Our results indicate that epinephrine, phenylephrine and methoxamine all act mainly via mixed subtypes of alpha(1)-adrenoceptors to induce cutaneous anesthesia in the rat.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U101
5-Methylurapidil, solid
C21H31N5O3