EMAIL THIS PAGE TO A FRIEND
Traffic (Copenhagen, Denmark)
Stonins--specialized adaptors for synaptic vesicle recycling and beyond?
PMID 19732400
Abstract
Stonins are a small family of evolutionarily conserved clathrin adaptor complex AP-2mu-related factors that may act as cargo-specific sorting adaptors in endocytosis and perhaps beyond. Whereas little is known about the localization and function of stonin 1, recent work suggests that stonin 2 serves as a linker between the endocytic proteins AP-2 and Eps15 and the calcium-sensing synaptic vesicle (SV) protein synaptotagmin 1. The molecular determinants involved in the recognition of SV cargo by the mu-homology domain of stonin 2 are evolutionarily conserved from worm to man, thereby identifying stonin 2 and its invertebrate homologs uncoordinated (UNC)-41 and stoned B as endocytic adaptors dedicated to the retrieval of surface-stranded SV proteins, most notably synaptotagmin. In this review, we summarize the current state of knowledge about mammalian stonins with a special focus on the role of stonin 2 in SV recycling at presynaptic nerve terminals.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
AV31514
Anti-SALF antibody produced in rabbit, IgG fraction of antiserum