EMAIL THIS PAGE TO A FRIEND
Natural product research
A new fungicidal lactone from Xylocarpus granatum (Meliaceae).
PMID 19735046
Abstract
A new lactone was isolated from the leaves of Xylocarpus granatum, along with three known compounds: triacontanol, beta-sitosterol and kaempferol-3-O-beta-D-glucoside. Its structure was elucidated as 3-(1-hydroxyethyl)-4,4-dimethyl-4-butyrolactone (1) by infrared, (1)H and (13)C NMR and ESI-MS data. At a concentration of 20 microg mL(-1), the new lactone gave a 67.4% inhibition rate against wheat powdery mildew.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
T3777
1-Triacontanol, ≥98% (capillary GC)
C30H62O