EMAIL THIS PAGE TO A FRIEND
Journal of hazardous materials
Modification of chloromethylated polystyrene with 2-mercabtobenzothiazole for application as a new sorbent for preconcentration and determination of Ag+ from different matrices.
PMID 19740598
Abstract
Chloromethylated polystyrene polymer (CMSP) modified with 2-mercabtobenzothiazole (MBT) has been developed for the selective separation and/or preconcentration of silver. The modified polymer (CMS-MBT) was characterized by elemental analysis and IR spectra. Batch and column modes were applied. The newly designed polymer quantitatively sorbed Ag(+) at pH 2 when the flow rate is 5 ml min(-1). The maximum sorption capacity was 0.493 mmol g(-1) while the preconcentration factor was 250 for Ag(+). The detection limit was 8 ng ml(-1). The desorption was effective with 5 ml of 2 mol l(-1) HNO(3) prior to detection using AAS. The modified polymer was highly ion-selective in nature even in the presence of large concentrations of electrolytes or organic media, with a preconcentrating ability for Ag(+). The utility of the modified polymer to synthetic and drugs samples showed RSD values of <3% reflecting its accuracy and reproducibility.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M3302
2-Mercaptobenzothiazole, 97%
C7H5NS2
63730
2-Mercaptobenzothiazole, technical, ≥90% (T)
C7H5NS2