EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
The effect of a hydroxy protecting group on the PtCl2-catalyzed cyclization of dienynes--a novel, efficient, and selective synthesis of carbocycles.
PMID 19746619
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
29598
Cyclooctane, analytical standard
C8H16
C109401
Cyclooctane, ≥99%
C8H16