EMAIL THIS PAGE TO A FRIEND
Pharmacology, biochemistry, and behavior
Effect of kappa-opioid receptor agonists U69593, U50488H, spiradoline and salvinorin A on cocaine-induced drug-seeking in rats.
PMID 19747933
Abstract
Our previous work indicated that pretreatment with the selective kappa-opioid receptor (KOPr) agonist, U69593, attenuated the ability of priming injections of cocaine to reinstate extinguished cocaine-seeking behavior. The present study expanded these initial tests to include other traditional KOPr agonists, U50488H, spiradoline (SPR), and salvinorin A (Sal A), an active constituent of the plant Salvia divinorum. Following acquisition and stabilization of cocaine self-administration, cocaine-produced drug-seeking was measured. This test was conducted in a single day and comprised an initial phase of self-administration, followed by a phase of extinguished responding. The final phase examined reinstatement of extinguished cocaine self-administration followed by a priming injection of cocaine (20.0mg/kg, intraperitoneal (I.P.)) in combination with the various KOPr agonists. Cocaine-induced drug-seeking was attenuated by pretreatment with U69593 (0.3mg/kg, subcutaneous (S.C.)), U50488H (30.0mg/kg, I.P.), SPR (1.0, 3.0mg/kg, I.P.) and Sal A (0.3, 1.0mg/kg, I.P.). Sal A (0.3, 1.0mg/kg, I.P.) had no effect on operant responding to obtain sucrose reinforcement or on cocaine-induced hyperactivity. These findings show that Sal A, like other traditional KOPr agonists attenuates cocaine-induced drug-seeking behavior.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
U105
U-62066, solid
C22H30Cl2N2O2 · CH4O3S
U103
U-69593, solid
C22H32N2O2