EMAIL THIS PAGE TO A FRIEND
Nucleic acids symposium series (2004)
Acridone-labeled DNA aptamer for the detection of biomolecules.
PMID 19749357
Abstract
An acridone-labeled DNA aptamer was synthesized to produce an aptamer probe. This aptamer probe could detect a specific molecule based on conformational changes upon binding of the specific molecules and the quenching of acridone emission.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
150215
9(10H)-Acridanone, 99%
C13H9NO
A3913
L-Argininamide dihydrochloride
C6H15N5O · 2HCl