EMAIL THIS PAGE TO A FRIEND
Angewandte Chemie (International ed. in English)
Stereocontrolled synthesis of the quinaldic acid macrocyclic system of thiostrepton.
PMID 19750639
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
160660
Quinaldic acid, 98%
C10H7NO2