EMAIL THIS PAGE TO A FRIEND
Biotechnology letters
Diastereoselective synthesis of L: -threo-3,4-dihydroxyphenylserine by low-specific L: -threonine aldolase mutants.
PMID 19760118
Abstract
Diastereoselectivity-enhanced mutants of L: -threonine aldolase (L: -TA) for L: -threo-3,4-dihydroxyphenylserine (L: -threo-DOPS) synthesis were isolated by error-prone PCR followed by a high-throughput screening. The most improved mutant was achieved from the mutant T3-3mm2, showing a 4-fold increase over the wild-type L: -TA. When aldol condensation activity was examined using whole cells of T3-3mm2, its de was constantly maintained at 55% during the batch reactions for 80 h, yielding 3.8 mg L: -threo-DOPS/ml.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9446
L-DOPS, ≥98% (HPLC)
C9H11NO5