EMAIL THIS PAGE TO A FRIEND
Organic & biomolecular chemistry
FtmOx1, a non-heme Fe(II) and alpha-ketoglutarate-dependent dioxygenase, catalyses the endoperoxide formation of verruculogen in Aspergillus fumigatus.
PMID 19763315
Abstract
Verruculogen is a tremorgenic mycotoxin and contains an endoperoxide bond. In this study, we describe the cloning, overexpression and purification of a non-heme Fe(ii) and alpha-ketoglutarate-dependent dioxygenase FtmOx1 from Aspergillus fumigatus, which catalyses the conversion of fumitremorgin B to verruculogen by inserting an endoperoxide bond between two prenyl moieties. Incubation with (18)O(2)-enriched atmosphere demonstrated that both oxygen atoms of the endoperoxide bond are derived from one molecule of O(2). FtmOx1 is the first endoperoxide-forming non-heme Fe(ii) and alpha-ketoglutarate-dependent dioxygenase reported so far. A mechanism of FtmOx1-catalysed verruculogen formation is postulated and discussed.