EMAIL THIS PAGE TO A FRIEND
Molecular diversity
One-pot synthesis of functionalized 4-oxo-2-thioxo-1,3-thiazinanes from primary amines, CS(2), and itaconic anhydride.
PMID 19763858
Abstract
An efficient synthesis of 2-(3-alkyl-4-oxo-2- thioxo-1,3-thiazinan-5-yl)acetic acids is described via a three-component reaction between primary amines, CS(2), and itaconic anhydride.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
I29204
Itaconic acid, ≥99%
C5H6O4