EMAIL THIS PAGE TO A FRIEND
Metabolism, Clinical and Experimental
Superoxide production by mitochondria of insulin-sensitive tissues: mechanistic differences and effect of early diabetes.
PMID 19765776
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
06-984
Anti-Mn-SOD Antibody, Upstate®, from rabbit