EMAIL THIS PAGE TO A FRIEND
Cell transplantation
Hepatocytes from fibrotic liver possess high growth potential in vivo.
PMID 19775529
Abstract
Hepatocyte transplantation is effective for treating liver failure, but healthy donors as a source of hepatocytes are quite limited. The livers of patients with hepatic fibrosis could be an alternative source; however, few reports have examined the nature of hepatocytes from fibrotic livers (f-hepatocytes). In this study, we compared the growth of f-hepatocytes and hepatocytes from normal livers (n-hepatocytes). Hepatocytes were isolated from normal and CCl(4)-treated wild-type Fischer rats that express dipeptidyl dipeptidase IV (DPPIV) gene (DPPIV(+)). The n- and f-hepatocytes proliferated in culture at similar rates. Both types of hepatocytes were transplanted into DPPIV(-) mutant Fischer rats that had been treated with retrorsine to injure the liver and were partially hepatectomized (PHx) before transplantation. Both n- and f-DPPIV(+)-hepatocytes proliferated and formed colonies. The colony sizes of f-hepatocytes 21 days posttransplantation were approximately three times those of n-hepatocytes. The hepatocytes were analyzed using a fluorescence activated cell sorter (FACS). The FACS profile differed between f- and n-hepatocytes: f-hepatocytes were less granular, less autofluorescent, and smaller than n-hepatocytes. These characteristics of f-hepatocytes resembled those reported for small-sized n-hepatocytes (SHs), which are highly proliferative and preferentially express a unique set of 10 SH genes. However, f-hepatocytes preferentially expressed only five of the SH genes. The expression profile of f-hepatocytes was rather similar to that of proliferating n-hepatocytes in the regenerating liver after PHx. The f-hepatocytes were morphologically normal and did not show any preneoplastic phenotype. These normal and proliferative natures of f-hepatocytes in vivo suggest the fibrotic liver as a source of hepatocytes for transplantation.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R0382
Retrorsine, ≥90% (HPLC)
C18H25NO6
37025
Retrorsine, analytical standard
C18H25NO6