EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Protecting-group-free total synthesis of (+/-)-subincanadine F.
PMID 19778081
Abstract
With chemoselective Dieckmann condensation as the key step, the protective-group-free total synthesis of (+/-)-subincanadine F was accomplished in 7 steps from the commercially available tryptamine in 33% overall yield.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
16491
Methyl bromopyruvate, technical
C4H5BrO3