EMAIL THIS PAGE TO A FRIEND
American journal of kidney diseases : the official journal of the National Kidney Foundation
Quiz page October 2009. Tetramine poisoning. Neurologic symptoms in a dialysis patient after ingesting seafood.
PMID 19781449