EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
Neuronal discharge hypersynchrony and the intracranial water balance in relation to glutamic acid and taurine redistribution: migraine and epilepsy.
PMID 1978346