EMAIL THIS PAGE TO A FRIEND
Molecules (Basel, Switzerland)
Cesium carbonate-catalyzed alpha-phenylchalcogenation of carbonyl compounds with diphenyl dichalcogenide.
PMID 19783931
Abstract
It was found that cesium carbonate has a unique catalytic ability on the reaction of carbonyl compounds with diphenyl diselenide to give the corresponding alpha-phenylseleno carbonyl compounds in moderate to good yields. Similarly, the alpha-phenylthiolation of carbonyl compounds with diphenyl disulfide was promoted by the cesium carbonate catalyst.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
169021
Phenyl disulfide, 99%
C12H10S2