EMAIL THIS PAGE TO A FRIEND
Analytical chemistry
Revision of analytical conditions for determining ligand molecules specific to soft metal ions using dequenching of copper(I)-bathocuproine disulfonate as a detection system.
PMID 19785444
Abstract
Reoptimization of analytical conditions was performed for a high-performance liquid chromatographic (HPLC) detection system for Cu(I) chelators based on the dequenching of Cu(I)-bathocuproine disulfonate complexes that occurs in the presence of Cu(I) chelators. The revision corrects for emission and excitation wavelengths that were in fact second-order light of the actual optimal wavelengths and for the composition of the postcolumn solution. These revisions resulted in an order of magnitude decrease in detection limits of phytochelatins, a class of cysteine-rich, heavy metal-binding peptides. The revised technique is capable of phytochelatin quantitation at femtomole quantities.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B1125
Bathocuproinedisulfonic acid disodium salt, for spectrophotometric det. of Cu, Fe
C26H18N2Na2O6S2