Analytica chimica acta
Synergetic enhancement effect of ionic liquid and diethyldithiocarbamate on the chemical vapor generation of nickel for its atomic fluorescence spectrometric determination in biological samples.
PMID 19786174
Room-temperature ionic liquid in combination with sodium diethyldithiocarbamate (DDTC) was used to synergetically improve the chemical vapor generation (CVG) of nickel. Volatile species of nickel were effectively generated through reduction of acidified analyte solution with KBH(4) in the presence of 0.02% DDTC and 25mmolL(-1) 1-butyl-3-methylimidazolium bromide ([C(4)mim]Br) at room temperature. Thus, a new flow injection (FI)-CVG-atomic fluorescence spectrometric (FI-CVG-AFS) method was developed for determination of nickel with a detection limit of 0.65microgL(-1) (3s) and a sampling frequency of 180h(-1). With consumption of 0.5mL sample solution, an enhancement factor of 2400 was obtained. The precision (RSD) for eleven replicate determinations of 20microgL(-1) Ni was 3.4%. The developed FI-CVG-AFS method was successfully applied to determination of trace Ni in several certified biological reference materials.