EMAIL THIS PAGE TO A FRIEND
Zeitschrift fur Naturforschung. C, Journal of biosciences
Bioactivity of flavonoids isolated from Lychnophora markgravii against Leishmania amazonensis amastigotes.
PMID 19791501
Abstract
The bioactivity of the flavonoids pinostrobin (1), pinocembrin (2), tectochrysin (3), galangin 3-methyl ether (4), and tiliroside (5) isolated from Lychnophora markgravii aerial parts was investigated in vitro against amastigote stages of Leishmania amazonensis. The compounds were isolated by several chromatographic techniques and their chemical structures were established by ESI-MS and NMR spectroscopic data. The flavonoids 1 and 3 were the most active compounds; they markedly reduced the viability of Leishmania amastigotes.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
79257
Tiliroside, analytical standard
C30H26O13