EMAIL THIS PAGE TO A FRIEND
Journal of ethnopharmacology
In vivo antioxidative activity of a quantified Pueraria lobata root extract.
PMID 19799984
Abstract
Oxidative stress has been associated with many pathological disorders such as atherosclerosis, diabetes and cancer. Supplementation with exogenous antioxidants, including phenolic compounds from plant sources, may help to restore the pro-oxidative/antioxidative balance. To take into account effects of absorption, metabolisation, plasma protein binding, distribution, and elimination, antioxidative research should not be limited to in vitro assays but be extended to in vivo models. In the present work a quantified 50% EtOH root extract of Pueraria lobata (Willd.) Ohwi (Fabaceae) was selected to determine its in vivo antioxidative activity in a diabetic rat model, where diabetes and the accompanying oxidative stress were induced by intraperitoneal administration of streptozotocin. This root extract was found to contain 10.42+/-0.15% puerarin as the main constituent and smaller amounts of the related isoflavonoids 3'-hydroxypuerarin, 3'-methoxypuerarin, 6''-xylosylpuerarin, daidzin, genistin, daidzein and genistein, as determined by a validated HPLC method. This extract was administered orally at a daily dose of 500 mg/kg root extract, corresponding to 50mg/kg puerarin, during 3 weeks. In addition the effect on the plasma concentration of some fat-soluble antioxidants (co-enzyme Q(9), alpha- and gamma-tocopherol) was evaluated. The level of malondialdehyde (MDA) in plasma, used as a marker of oxidative damage to lipids, was reduced to the same level as in healthy control animals, and as in the positive control group treated daily with 50mg/kg alpha-tocopherol acetate. No obvious signs of toxicity were observed by administration of 10x the treatment dose.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27597
Coenzyme Q9, ≥96.0% (HPLC)
C54H82O4