EMAIL THIS PAGE TO A FRIEND
Life sciences
Somatostatin content and binding in small intestinal mucosa from fed, fasted, and refed rabbits.
PMID 1980331
Abstract
The present study is an investigation of the effects of 12- to 96-hours' starvation and 96-hours' starvation plus 48-hours' refeeding on both somatostatin-like immunoreactivity (SLI) and cytosolic somatostatin binding sites in rabbit small intestinal mucosa. The SLI concentration increased after 24 h in duodenal and jejunal mucosa, but not in ileal mucosa, and reached its highest value after 96 h of fasting. The number of specific high and low-affinity somatostatin binding sites, but not their affinity, decreased with the duration of fasting in the same gut segments, refeeding of fasted animals resulted in a return to normal control values for small intestine mucosal SLI and somatostatin binding.