EMAIL THIS PAGE TO A FRIEND
Minerva anestesiologica
[Antagonism with flumazenil of sedation by benzodiazepine during loco-regional anesthesia].
PMID 1980354
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D7662
Delorazepam
C15H10Cl2N2O